missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Low Range Quantitative DNA Ladder
GREENER_CHOICE

Product Code. 10246832
Click to view available options
Quantity:
100 applications
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10246832

Brand: Invitrogen™ 12373031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

The E-Gel™ Low Range Quantitative DNA Ladder is supplied at 17.5ng/μL in 8mM Tris-HCl (pH 7.5), 1mM EDTA, 5% glycerol, and 0.005% Orange G; For 100 applications

Suitable for estimating size and quantity of double-stranded DNA by ethidium bromide visualization on all E-Gel™

Prepared from a specifically constructed plasmid, the ladder consists of an equimolar mixture of five blunt-ended fragments ranging from 100 to 2000bp. This ladder is supplied ready to use in an optimized E-Gel loading buffer. The buffer includes Orange G dye that runs at approximately 25bp. Compatible with E-Gel™, E-Gel™ 48, E-Gel™ 96 gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 mL E-Gel Low Range Quantitative DNA Ladder
• 1 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods, store at -20°C.
Concentration 0.0175 μg/μL
Gel Type Agarose
Product Type DNA Ladder
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 100 to 2000 bp
Volume (Metric) 1000 μL
Gel Compatibility E-Gel™ agarose gels
Green Features Sustainable packaging
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.