Learn More
Invitrogen™ Easy-DNA™ gDNA Purification Kit
The Easy-DNA™ Kit is fast and easy way to isolate high-quality, high-molecular weight genomic DNA (gDNA) from a wide range of cells and tissue types.
Brand: Invitrogen™ K180001
492.10 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.75000kg
Includes: The Easy-DNA Kit contains enough reagents for 15–200 samples, depending on sample size and type.This kit contains: 55mL Solution A (Lysis Solution), 25mL Solution B (Precipitation Solution), 100mL TE Buffer, 750μl Mussel Glycogen (2mg/mL in sterile water), 750 μl RNase (2mg/mL in sterile water), 750μl Protein Degrader (5mg/mL in sterile water)
Description
The Easy-DNA™ Kit is fast and easy way to isolate high-quality, high-molecular weight genomic DNA (gDNA) from a wide range of cells and tissue types. Sample sizes can range from single hair follicle up to 1 gram of mammalian tissue. Other samples tested include mouse tails, plant leaves, yeast, and E. coli cells. These samples yield high-quality DNA with average size between 100–200 kb, which is suitable for PCR, DNA hybridization, genomic DNA library construction, and other applications.
- Reliability: High-quality genomic DNA from large or small samples of cells, tissue, plant, yeast, E. coli, blood, and even hair follicles
- Simplicity: For most sample types, only 4 steps, typically requiring less than 90 minutes are needed to isolate high-quality DNA
- Versatility: One kit is supported by optimized protocols for variety of cell and tissue types
Order Info
Shipping Condition: Room Temperature
Specifications
• 55 mL Solution A; room temperature • 25 mL Solution B; room temperature • 100 mL TE Buffer; room temperature • 750 μL Mussel Glycogen; room temperature, or -20°C for long-term storage • 750 μL RNase; room temperature, or -20°C for long-term storage • 750 μL Protein Degrader; room temperature, or -20°C for long-term storage |
|
Bacteria, Cells, Hair Follicles, Mouse Tails, Plant, Tissue, Virus, Whole Blood, Yeast | |
gDNA | |
Not High-throughput Compatible (Manual) | |
Room Temperature | |
≤850 μg |
Organic Extraction | |
10 to 150 μL | |
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization | |
1 Kit | |
Up to 90 min. | |
Bacteria: ≤109 cells Cells: ≤108 Hair Follicles: ≤5 Mouse Tails: 1 cm Plant: ≤50 mg Tissue: ≤1 g Virus: ≤750 μL Whole Blood: ≤2 mL Yeast: ≤10 mL |
For Research Use Only. Not for use in diagnostic procedures.