Learn More
Invitrogen™ EveryPrep™ Universal Vacuum Manifold
The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids.
Brand: Invitrogen™ K211101
853.37 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : Weight : 6.96000kg
Description
The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids. It offers the ability to directly elute from the manifold using the new ChargeSwitch™ Pro Filter Plasmid Mini, Midi and Maxi Kits and also many other competitors' vacuum assisted spin column preps and filter plates. The EveryPrep™ Universal Vacuum Manifold replaces and consolidates all previous manifolds into one space saving benchtop unit. The manifold reduces sample handling to a minimum by allowing direct parallel and simultaneous processing of up to 96 samples using collection plates, deep-well blocks, or microtubes. The use of a vacuum eliminates the need for time-consuming centrifugation. The manifold will consist of a dual chamber construct fashioned out of aluminum with interchangeable top plates of a luer-lock tapped top plate, a 96-well top plate, a mini column elution plate and a maxi column elution plate.
Order Info
Shipping Condition: Room Temperature
Specifications
Vacuum Manifold | |
1 vacuum manifold. Store at room temperature. | |
1 Ea. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.