Learn More
Invitrogen™ Filter Cartridges & Tubes for cDNA Purification
For use with the DNAclear™ cDNA Purification Kit
Brand: Invitrogen™ AM10066G
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: Each package contains 10 cartridges and tubes.
Description
The DNAclear cDNA Purification Kit is designed to purify double-stranded DNA (dsDNA) generated by enzymatic reactions such as cDNA synthesis and PCR.
The process is simple and fast. DNA amounts ranging from 1ng to 3μg can be efficiently purified and concentrated. The DNAclear procedure consists of three steps: 1) DNA is bound to the Microfilter Cartridge in the presence of the DNA Binding Buffer, 2) the DNA is washed with the cDNA Wash Buffer, and 3) the DNA is eluted with preheated Nuclease-free Water. The DNAclear Kit can be used to remove free nucleotides, oligonucleotides, proteins, and salts from dsDNA samples. The DNA that is recovered from the Microfilter Cartridge is ready for any application that requires highly pure DNA including in vitro transcription reactions. The DNAclear Kit includes enough materials to perform 20 purifications.
- Efficiently purify and concentrate dsDNA ranging from 1ng to 3μg
- Ideal for downstream applications such as in vitro transcription
- Simple and fast protocol – about 10 minutes
Cloning, DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up, cDNA Libraries and Library Construction
Order Info
Shipping Condition: Room Temperature
Specifications
Filter Cartridge and Tube | |
PCR Products, cDNA | |
Tube, Spin Column | |
10 Ea. |
Each package contains 10 cartridges and tubes. | |
Room Temperature | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.