missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flp-In™-293 Cell Line

Product Code. 10164282
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10164282

Brand: Invitrogen™ R75007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

1 x 107 cells are supplied frozen in 1mL of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen.

Cells contain a single stably integrated FRT site at a transcriptionally active genomic locus

Flp-In 293 cell line is designed for rapid generation of stable cell lines that ensure high level expression of your protein of interest from a Flp-In expression vector. Targeted integration of a Flp-In expression vector ensures high-level expression of your gene of interest. Flp-In-293 cell line was created by transfecting the parent cell lines with pFRT/lacZeo and selecting for stable Zeocin™- resistant clones. Flp-In-293 cells work well with Flp-In vectors that express a gene from the CMV promoter (e.g., pcDNA™5/FRT, pcDNA5/FRT/V5-His-TOPO™, and pSecTag/FRT/V5-His-TOPO

Constitutive Expression, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 10E7 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.
Cell Line Flp-In-293
Species Human
No. of Cells 1 mL
Product Line Flp-In
Quantity 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.