Learn More
Invitrogen™ Flp-In™-CHO Cell Line
Designed for rapid generation of stable cell lines that express protein of interest from Flp-In expression vector
Brand: Invitrogen™ R75807
1353.20 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Includes: 3 x 106 cells are supplied frozen in 1mL 90% complete medium and 10% DMSO.
Description
- These cells contain single stably integrated FRT site at transcriptionally active genomic locus
- Targeted integration of Flp-Inexpression vector ensures high-level expression of gene of interest
- Created by transfecting parent cell lines with pFRT/lacZeo and selecting for stable Zeocin™-resistant clones
- The Flp-In-CHO Cell Line was created by transfecting CHO cells with pFRT/lacZeo2 and selecting for Zeocin™-resistant clones
- Flp-In-CHO cell lines work well with Flp-In vectors that express gene from CMV promoter (pcDNA™5/FRT, pcDNA5/FRT/V5-His-TOPO™, and pSecTag/FRT/V5-His-TOPO™)
Constitutive Expression, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration
Specifications
3 x 106 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. | |
Hamster | |
Flp-In™ |
Flp-In™-CHO Cell Line | |
pSec |
For Research Use Only. Not for use in diagnostic procedures.