Learn More
Invitrogen™ Flp-In™-Jurkat Cell Line
Designed for rapid generation of stable cell lines that express protein of interest from Flp-In expression vector
Brand: Invitrogen™ R76207
Code : 50
Additional Details : Weight : 1.25000kg
Includes: 3 x 106 cells are supplied frozen in 1mL 90% complete medium and 10% DMSO.
Description
- These cells contain single stably integrated FRT site at transcriptionally active genomic locus
- Targeted integration of Flp-Inexpression vector ensures high-level expression of gene of interest
- Created by transfecting the parent cell lines with pFRT/lacZeo and selecting for stable Zeocin™-resistant clones
- Co-transfection of Flp-InCell Lines with Flp-Inexpression vector and Flp recombinase vector, pOG44, results in targeted integration of expression vector to same locus in every cell, ensuring homogeneous levels of gene expression
- Flp-In-Jurkat cell lines work well with vectors that express gene from CMV promoter
Constitutive Expression, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration
Specifications
3 x 106 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. | |
Human | |
Flp-In™ |
Flp-In™-Jurkat Cell Line | |
pSec |
For Research Use Only. Not for use in diagnostic procedures.