Learn More
Invitrogen™ β-Gal Assay Kit
Provides reagents required to quickly measure levels of active β-galactosidase expressed in cells transfected with plasmids expressing lacZgene
Brand: Invitrogen™ K145501
351.05 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg Transport : UN number : 2966 Chem class : 6.1 Pack group : II
Includes: 10X PBS (phosphate buffered saline), lysis buffer, cleavage buffer, β-mercaptoethanol, ONPG, stop buffer.
Description
- lacZis bacterial gene often used as reporter construct in eukaryotic transfection experiments
- Gene product of lacZ, β-galactosidase, is resistant to proteolysis in cellular lysates, and its activity is easily assayed
- β-galactosidase catalyzes hydrolysis of β-galactosidase such as ortho-nitrophenyl-D-galactopyranoside (ONPG)
- Hydrolysis of ONPG to ONP anion produces bright yellow color with peak absorbance at 420nm that can be quantified using spectrophotometer
- 10.7 gene expression
- Colorimetric detection
Beta-Galactosidase Assays, Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifications
The β-Gal Assay Kit contains 10X PBS (phosphate buffered saline), lysis buffer, cleavage buffer, β-mercaptoethanol, ONPG, stop buffer. Store ONPG at -20°C. All other components are stored at either room temperature or +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
Colorimetric | |
ONPG |
β-Gal (lacZ) Assay | |
Beta-Gal Assay Kit | |
Beta-Galactosidase |
For Research Use Only. Not for use in diagnostic procedures.