Learn More
Invitrogen™ Champion™ pET300/NT-DEST and pET301/CT-DEST Gateway™ Vector Kit
Designed for rapid cloning with Gateway entry clone and subsequent high-level prokaryotic expression controlled by strong bacteriophage T7 promoter
Brand: Invitrogen™ K630001
Code : 50
Additional Details : Weight : 0.02000kg
Description
- In addition to T7 promoter, each vector contains only necessary functional elements and N- or C-terminal 6xHis tag (pET300/NT-DEST and pET301/CT-DEST, respectively) for convenient purification and detection
- The vector kit is excellent for structural biologists who desire no or minimal modifications to their protein of interest
Bacterial Expression, Bioproduction, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
T7 | |
None | |
His Tag (6x) | |
pET, pDEST |
IPTG | |
DNA Cloning Vector Kit | |
Ampicillin (AmpR) | |
Gateway | |
Champion™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.