Learn More
Invitrogen™ pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ Mach1™-T1R E. coli and PureLink™ Kit
Product Code. 10369892
Click to view available options
Quantity:
20 Preps
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10369892
Brand: Invitrogen™ K252002
For Research Use Only. All usage must comply with product instructions.
Includes
pCR8/GW/TOPO Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers; TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes.
Delivers simple, 5-minute cloning and access to Gateway recombination cloning vector platform
  • pCR™8 TOPO TA vector contains minimal MCS, so sequence more of insert and less of vector
  • Patented att sites make pCR™8 vector is simplest way to create an entry clone, which facilitates easy access via Gateway recombination cloning to variety of expression vectors without tedious sub-cloning, saving valuable research time
  • pCR™8/GW/TOPO TA Cloning Kit features fast growing E. coli that shortens cloning time
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • Sequencing primer sites within 55 bp of insertion site
  • Spectinomycin resistance gene for robust selection in E. coli
  • EcoR I sites flanking the PCR product insertion site for easy excision of inserts
Cloning,Gateway Cloning,PCR Cloning
TRUSTED_SUSTAINABILITY
Specifications
Cloning Method TOPO™ with Gateway™
Content And Storage pCR™8/GW/TOPO™ Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers; One Shot™ TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink™ Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes. Store all components of the PureLink™ kit at room temperature. Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
No. of Reactions 20 preps
Product Line One Shot
Product Type TA Cloning Kit
Quantity 20 Preps
Vector pCR