missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ TOP10 E. coli

Product Code. 10532893
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10532893

Brand: Invitrogen™ K250020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

TOPO-adapted, ready-to-use pCR8/GW/TOPO Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers, TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid.

Delivers simple, 5-minute cloning and access to Gateway recombination cloning vector platform

  • Contains minimal MCS, so sequence more of insert and less of vector
  • Patented att sites, pCR™8 vector to create entry clone, which facilitates easy access via Gateway recombination cloning to variety of expression vectors without tedious sub-cloning, saving valuable research time
  • att sites for rapid recombination into a variety of Gateway destination vectors
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • Sequencing primer sites within 55 bp of insertion site
  • Spectinomycin resistance gene for robust selection in E. coli
  • EcoRI sites flanking the PCR product insertion site for easy excision of inserts

Cloning,Gateway Cloning,PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Cloning Method TOPO™ with Gateway™
Content And Storage The pCR™8/GW/TOPO™ TA Cloning Kit contains the TOPO™-adapted, ready-to-use pCR™8/GW/TOPO™ Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers, One Shot™ TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
No. of Reactions 20 reactions
Product Line One Shot
Product Type TA Cloning Kit
Quantity 20 Reactions
Vector pCR

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.