missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ pDONR™221 Vector

Product Code. 11568706
Click to view available options
Quantity:
6 μg
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11568706

Brand: Invitrogen™ 12536017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

6μg pDONR221 vector, lyophilized in TE buffer, pH 8.0.

Enables rapid cloning of one or more genes into virtually any protein expression system

  • Designed to create entry clone following PCR with attB-modified custom primers
  • With entry clone, recombine gene of interest into variety of expression vectors adapted for use in Gateway technology
  • PCR product is directionally cloned with efficiencies typically >99%
  • Rapid, efficient, directional cloning of PCR products
  • Versatility to use any amplification enzyme
  • Cloning without need for restriction endonucleases or ligase
  • Selection of entry clones with kanamycin or Zeocin™
  • In addition, pDONR221 and pDONR™/Zeo vectors have pUC origin for high plasmid yields and universal M13 sequencing sites for ease of use

Cloning, Gateway Cloning

TRUSTED_SUSTAINABILITY

Specifications

Product Type Gateway System Destination Expression Vector
Content And Storage 6 μg pDONR221™ vector, lyophilized in TE buffer, pH 8.0. Upon receipt, store at -20°C.
Cloning Method Gateway
Quantity 6 μg
Vector pDONR
Product Line Gateway, pDONR

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.