Learn More
Invitrogen™ Gateway™ pDONR™/Zeo Vector
Product Code. 11558706
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
For Research Use Only. All usage must comply with product instructions.
Product Code. 11558706
Brand: Invitrogen™ 12535035
For Research Use Only. All usage must comply with product instructions.
Includes
6μg pDONR/Zeo vector, lyophilized in TE buffer, pH 8.0; 1.25mL Zeocin. Zeocin is provided as a 100mg/mL solution in deionized, sterile water.
Enables rapid cloning of one or more genes into virtually any protein expression system
  • Designed to create entry clone following PCR with attB-modified custom primers
  • With entry clone, recombine gene of interest into variety of expression vectors adapted for use in Gateway technology
  • PCR product is directionally cloned with efficiencies typically >99%
  • pDONR/Zeo vector has pUC origin for high plasmid yields and universal M13 sequencing sites for ease of use
Cloning, Gateway Cloning
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gateway System Destination Expression Vector
Content And Storage 6 μg pDONR™/Zeo vector, lyophilized in TE buffer, pH 8.0. pDONR™/Zeo is also supplied with 1.25 ml Zeocin™. Zeocin™ is provided as a 100 mg/ml solution in deionized, sterile water. Upon receipt, store the pDONR™/Zeo vector at -20°C and the Zeocin™ at -20°C, protected from light.
Cloning Method Gateway
Quantity 6 μg
Vector pDONR
Product Line Gateway, pDONR