Learn More
Invitrogen™ pTrcHis A, B, & C Bacterial Expression Vectors
Offer enhanced translation initiation and high-level expression in E. coli
Brand: Invitrogen™ V36020
Code : 50
Additional Details : Weight : 0.06000kg
Includes: All vectors are provided in suspension. A TOP10 E. coli stab and positive expression control is also provided.
Description
- These vectors feature high-level regulated transcription from the trc promoter
- Enhanced translation efficiency of eukaryotic genes in E. coli
- The lacO operator and lacIq repressor gene for transcriptional regulation in any E. coli strain
- This particular vector offers N-terminal polyhistidine (6xHis) tag for rapid purification with nickel-chelating resin and detection with an anti-HisG antibody
- N-terminal Xpress epitope for easy detection with an anti-Xpress antibody
- Enterokinase cleavage site for removal of fusion tag
Bacterial Expression, Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
Trc, lacO | |
None | |
Ampicillin (AmpR) | |
Thioredoxin | |
pTrc |
IPTG | |
Bacterial Expression Vector | |
All vectors are provided in suspension. A TOP10 E. coli stab and positive expression control is also provided. Store vectors at -20°C. Store stab at room temperature. The vector is guaranteed stable for 6 months when properly stored. | |
EK (Enterokinase) Recognition Site | |
Restriction Enzyme/MCS |
For Research Use Only. Not for use in diagnostic procedures.