Learn More
Invitrogen™ Vivid Colors™ pcDNA™6.2/C-YFP-DEST Vector
Allow quick and easy fusion of a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria
Brand: Invitrogen™ V35720
Code : 50
Additional Details : Weight : 0.00525kg
Includes: Each vector is supplied as supercoiled and lyophilized plasmid DNA.
Description
- Contain the Yellow Fluorescent Protein (YFP) for simple, noninvasive detection of recombinant protein
- Fluorescent proteins have been humanized for optimal mammalian expression
- Features the CMV promoter for high-level expression of the recombinant fluorescent fusion protein
- Fuse to the C-terminus of the protein
Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Cloning, Constitutive Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifications
Constitutive | |
CMV | |
YFP | |
Each vector is supplied as supercoiled and lyophilized plasmid DNA. Store at -20°C after resuspended. Guaranteed stable for 6 months when properly stored. | |
Gateway | |
Gateway™, Vivid Colors™, pcDNA™ |
Transfection | |
Mammalian Expression Vector | |
Blasticidin | |
YFP | |
pDEST, pcDNA | |
Reporter Assays, Subcellular Localization |
For Research Use Only. Not for use in diagnostic procedures.