Learn More
Invitrogen™ Vivid Colors™ pcDNA™6.2/N-EmGFP-DEST Vector
Allow quick and easy fusion of a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria
Brand: Invitrogen™ V35620
Code : 50
Additional Details : Weight : 0.00525kg
Includes: Each vector is supplied as supercoiled and lyophilized plasmid DNA.
Description
- These vectors contain the next-generation EGFP, Emerald Green Fluorescent Protein (EmGFP) for simple, non-invasive detection of recombinant protein
- Fluorescent proteins have been humanized for optimal mammalian expression
- Vectors offe the CMV promoter for high-level expression of the recombinant fluorescent fusion protein
- Fuse to the N-terminus of the protein
Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Cloning, Constitutive Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifications
Constitutive | |
CMV | |
GFP (EmGFP) | |
Each vector is supplied as supercoiled and lyophilized plasmid DNA. Store at -20°C after resuspended. Guaranteed stable for 6 months when properly stored. | |
Gateway | |
Gateway™, Vivid Colors™, pcDNA™ |
Transfection | |
Mammalian Expression Vector | |
Blasticidin | |
V5 Epitope Tag, GFP (EmGFP) | |
pDEST, pcDNA | |
Reporter Assays, Subcellular Localization |
For Research Use Only. Not for use in diagnostic procedures.