Learn More
Invitrogen™ Homozygous Diploid Complete Set
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Brand:  Invitrogen™ 95401.H1R3
Product Code. 10410065
  • £18280.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)