Learn More
Invitrogen™ Human Cot-1 DNA™-Fluorometric QC
Quantifying by fluorometry provides greater data accuracy
Brand: Invitrogen™ 15279101
Code : 50
Additional Details : Weight : 0.00525kg
Includes: Human Cot-1 DNA™-Fluorometric QC is supplied at 1mg/mL in 10mM Tris-HCl (pH 7.4), 1mM EDTA.
Description
Human Cot-1 DNA-Fluorometric QC is placental DNA predominantly 50 to 300 bp in size and enriched for repetitive DNA sequences, such as the Alu and Kpn family members (1,2). Quantifying Cot-1 DNA™ by fluorometry provides more accurate concentration data of higher molecular weight products, making it beneficial to certain genomic-based array experiments, such as array CGH using BAC arrays. Human Cot-1 DNA™-Fluorometric QC suppresses prevalent repetitive elements during hybridization, improving results.
Array CGH, Cell Analysis, Cellular Imaging, ChIP-on-Chip, Chromatin Biology, Fluorescence In Situ Hybridization, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, In Situ Hybridization (ISH), Microarray Analysis, RNAi, Epigenetics and Non-Coding RNA Research
Order Info
Shipping Condition: Dry Ice
Specifications
DNA QC | |
Cot-1 DNA | |
Human | |
Block Non-Specific Hybridization |
Human Cot-1 DNA™-Fluorometric QC is supplied at 1 mg/ml in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA. Store at -20°C. Guaranteed stable for 12 months when properly stored. | |
Dry Ice | |
Cot-1 DNA™ | |
1 mg |
For Research Use Only. Not for use in diagnostic procedures.