missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human miR RNAi Select
Easily design effective RNAi molecules for reliable results
Brand: Invitrogen™ 100002318
This item is not returnable.
View return policy
Description
- Using BLOCK-iT™ RNAi Designer, design Custom BLOCK-iT siRNA sequences for efficient and specific gene expression knockdowns in any organism
- For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Human miR RNAi Select | |
Human |
siRNA | |
1 Each |