missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Drosophila S2 Cells in Schneider's Medium

Derived from a primary culture of late stage (20-24 hours old) Drosophila melanogaster embryos

Brand:  Invitrogen™ R69007

429.30 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10412072

  • £530.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

Review the Material Safety Data Sheet before handling.

This item is not returnable. View return policy

Description

Description

Gibco Drosophila S2 cells are used for heterologous protein expression in the DrosophilaM Expression System (DES™). Many features of the S2 cell line suggest that it is derived from a macrophage-like lineage. S2 cells grow at room temperature in ambient CO2 as a loose, semi-adherent monolayer in tissue culture flasks and in suspension in spinners and shake flasks.

  • Expression of protein transiently or through generation of stable cell lines
  • Quality and performance testing

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen
Insect Cells
Drosophila S2 Cells
S2 (Schneider 2)
Drosophila
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.