Learn More
Invitrogen™ Pichia EasyComp™ Transformation Kit
Designed for rapid preparation and transformation of competent Pichia pastoris cells
Brand: Invitrogen™ K173001
403.75 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Includes: Sufficient reagents (Solutions I, II, and III) for 6 preparations of competent cells. Each competent cell preparation yields enough cells for 20 transformations.
Description
- Competent cells can be frozen for future use, minimizing waste
- Competent cells can be prepared in less than 30 minutes
- Competent cell preparation and transformation solutions are optimized for use with Pichia pastoris are provided ready to use and are tested to ensure results
Cloning, Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis/Transformation/Yeast Expression
Specifications
Pichia Transformation Kit | |
The Pichia EasyComp™ Transformation Kit contains sufficient reagents (Solutions I, II, and III) for 6 preparations of competent cells. Each competent cell preparation yields enough cells for 20 transformations. Store Solutions I and III at +4°C. Store Solution II at room temperature. All components are guaranteed stable for 6 months when properly stored. | |
P. pastoris | |
EasyComp™ |
For Research Use Only. Not for use in diagnostic procedures.