Learn More
Invitrogen™ Taq DNA Polymerase, recombinant
Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.
£89.50 - £2855.00
Specifications
Format | Stand-alone enzyme |
---|---|
GC-Rich PCR Performance | Low |
PCR Method | qPCR, Standard PCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Includes: Taq DNA Polymerase (5U/μL); 10X PCR buffer (200mM Tris-HCl pH 8.4, 500mM KCl); Magnesium Chloride (50mM)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10174762
|
Invitrogen™
10342053 |
100 Reactions |
£89.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10033382
|
Invitrogen™
10342020 |
500 Reactions |
£207.00
500 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11508626
|
Invitrogen™
10342046 |
1500 Reactions |
£592.00
1000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10063262
|
Invitrogen™
10342178 |
5000 Reactions |
£2855.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.
Features
- A choice of recombinant or native enzyme
- Amplification of PCR products up to 5 kb in size
- Enzyme is licensed and qualified for PCR
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- RT-PCR
- Sequencing ssDNA
- Cycle sequencing
Notes
- One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
- For superior PCR performance, DreamTaq DNA Polymerase is recommended.
Order Info
Shipping Condition: Approved for shipment on wet or dry ice
Specifications
Stand-alone enzyme | |
qPCR, Standard PCR | |
Standard | |
DNA Polymerase | |
Standard PCR | |
1X | |
3'-A | |
5 kb or less |
Low | |
Taq DNA Polymerase | |
5' - 3' | |
Dry Ice | |
5 U/μL | |
No | |
Separate Components | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.