missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBlot™ Lid Latch Replacement
Click to view available options
Type:
Lid Latch Replacement
Unit Size:
Each
Description
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | iBlot |
| Product Line | iBlot |
| Type | Lid Latch Replacement |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction