Learn More
Invitrogen™ INVSc1 S. cerevisiae Yeast Strain
Click to view available options
Quantity:
1 Stab
Unit Size:
Each
Description
The S. cerevisiae strain INVSc1 is a fast-growing strain ideal for expression. INVSc1 has the following genotype:
INVSc1: MATa his3D1 leu2 trp1-289 ura3-52 MAT his3D1 leu2 trp1-289 ura3-52
Specifications
Specifications
| Product Type | Yeast Strain |
| Bacterial or Yeast Strain | INVSc1 |
| Cell Line | Yeast |
| Species | S. cerevisiae |
| Quantity | 1 Stab |