Learn More
Invitrogen™ IPTG
IPTG (isopropylthio-β-galactoside), a structural non-metabolizable analogue of allolactose, is an inducer of β-galactosidase activity in bacteria and is suitable for use with X-Gal or Bluo-Gal to detect lac gene activity in cloning procedures.
Brand:  Invitrogen™ 15529019
43.70 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10397642
  • £57.50 / 1g
Edge
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
IPTG (isopropylthio-β-galactoside), a structural non-metabolizable analogue of allolactose, is an inducer of β-galactosidase activity in bacteria and is suitable for use with X-Gal or Bluo-Gal to detect lac gene activity in cloning procedures. IPTG binds to the lac repressor, releasing it from the lac operator, and thereby allowing the transcription of genes in the lac operon, such as the gene coding for β-galactosidase.
In addition, IPTG is one of the most efficient molecular inducers for promoter’s transcriptional activity and is used extensively for heterologous protein expression induction in E. coli. Though mainly used in cell culture assays, publications demonstrated the use IPTG in vivo in animal models too.
Primary applications
• Blue-white screening—induce functional β-galactosidase
• Induce protein expression from inducible T7 RNA polymerase E. coli expression systems egulated by lac operon
Find the media format that fits your needs
• We offer a wide variety of ready-to-use media formulations, such as LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).
• X-Gal (Cat. No. 15520018) or Bluo-Gal (Cat. No. 15519028) for blue-white screening.
Order Info
Approved for shipment at Room Temperature or on Wet Ice.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IPTG (Isopropyl β-D-thiogalactopyranoside)
Bottle
Approved for shipment at Room Temperature or on Wet Ice
• IPTG Powder (1 g)
Store at –20°C.
1 g
Powder