Learn More
Invitrogen™ Lipofectamine™ 2000 Transfection Reagent

Description
Lipofectamine™ 2000 Transfection Reagent is versatile transfection reagent that has been shown to effectively transfect widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine 2000 Transfection Reagent is best transfection reagent available from Life Technologies for the co-transfection of siRNA and plasmid DNA.
- Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression
- Superior performance for co-transfection of siRNA and plasmid DNA
- Proven efficacy in the presence of serum, eliminates the need to change media following transfection
- Reliable performance for high-throughput applications
- The best choice for establishing stable cell lines
Order Info
Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.
Specifications
Specifications
| Content And Storage | Contains one bottle (15 mL) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze. |
| Cell Type | Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR) |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| Green Features | Sustainable packaging |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | Lipofectamine |
| Product Type | Transfection Reagent |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.