missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
GREENER_CHOICE

Product Code. 10696153
missing translation for 'orderingAttributeHoverText'
Quantity:
0.3 mL
0.75 mL
1.5 mL
15 mL
missing translation for 'unitSize'
1.5mL
750µL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10696153

missing translation for 'mfr': Invitrogen™ 11668019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines

Lipofectamine™ 2000 Transfection Reagent is versatile transfection reagent that has been shown to effectively transfect widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine 2000 Transfection Reagent is best transfection reagent available from Life Technologies for the co-transfection of siRNA and plasmid DNA.

  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum, eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines

Order Info

Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one vial (1.5 ml) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Green Features Sustainable packaging
High-throughput Compatibility High-throughput Compatible
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 1.5 mL
Serum Compatible Yes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.