missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
GREENER_CHOICE

Product Code. 10696343
Click to view available options
Quantity:
0.3 mL
0.75 mL
1.5 mL
15 mL
Unit Size:
1.5mL
750µL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10696343

Brand: Invitrogen™ 11668027

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines

Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine™ 2000 Transfection Reagent is our best transfection reagent available for the co-transfection of siRNA and plasmid DNA.

With Lipofectamine™ 2000 Transfection Reagent, you'll get:

  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression (see table)
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum—eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines

    A high-performance transfection reagent for gene expression and gene silencing
    Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to achieve convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.

    Ideal for high-throughput work
    Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
  • Order Info

    Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.

    TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains one vial (0.75 ml) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze.
    Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
    Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
    Transfection Technique Lipid-based Transfection
    For Use With (Application) Transfection
    Green Features Sustainable packaging
    High-throughput Compatibility High-throughput Compatible
    Product Line Lipofectamine
    Product Type Transfection Reagent
    Quantity 0.75 mL
    Serum Compatible Yes
    Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.