missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent
Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments
£123.00 - £10150.00
Specifications
Cell Type | Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
---|---|
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Transfection Technique | Lipid-based Transfection |
For Use With (Application) | Transfection |
Green Features | Sustainable packaging |
Includes: Contains one vial (0.75mL) Lipofectamine RNAiMAX Transfection Reagent.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10514953
|
Invitrogen™
13778075 |
0.75 mL |
£612.00
750µL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10601435
|
Invitrogen™
13778150 |
1.5 mL |
£1008.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12333563
|
Invitrogen™
13778100 |
0.1 mL |
£123.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12323563
|
Invitrogen™
13778030 |
0.3 mL |
£376.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12343563
|
Invitrogen™
13778500 |
15 mL |
£10150.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.With Lipofectamine™ RNAiMAX Transfection Reagent you will get:
High knockdown in a wide range of cells
Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.
Simple, high-throughput–ready transfections
Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.
Bestellinformation
Shipping Condition: Qualified for Ambient or Wet Ice
Spezifikation
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Lipid-based Transfection | |
Sustainable packaging | |
Lipofectamine™ | |
Yes |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.