missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ Transfection Reagent

Product Code. 10615013
Click to view available options
Quantity:
0.3 mL
1 mL
4 mL
Unit Size:
1mL
Each
Pack of 4
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10615013

Brand: Invitrogen™ 18324020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains one vial of 1mL.

Lipofectamine™ Transfection Reagent is one of our first-generation, cationic-lipid transfection reagents formulated for the transfection of DNA into eukaryotic cells.

Lipofectamine™ Transfection Reagent is one of our first-generation, cationic-lipid transfection reagents formulated for the transfection of DNA into eukaryotic cells. Lipofectamine™ Transfection Reagent is a trusted, value reagent with many years of cited use in thousands of publications and with many cell lines.

With Lipofectamine™ Transfection Reagent, you'll get:

  • A reagent proven to work in high-throughput applications
  • A reliable reagent for establishing stable cell lines
  • A reagent that works well with PLUS™ reagent for higher protein expression

A proven technology

Our Lipofectamine™-brand reagents have been recognized as the most-cited transfection reagent family, with tens of thousands of citations since launch in 1993. Lipofectamine™ Transfection Reagent is no exception as a trusted reagent for a wide range of cell lines. When used in combination with PLUS™ Reagent, Lipofectamine™ Transfection Reagent has been shown to work well with cells such as BHK-21, NIH 3T3, COS-1, fibroblasts, keratinocytes, HT-29, MRC-5, and SK-BR3.

A 'value' alternative for plasmid DNA

With such a legacy comes a commitment to providing the best reagents for all scientists and labs, regardless of their funding capabilities. Lipofectamine™ Transfection Reagent is a value alternative for transfection of plasmid DNA into eukaryotic cells with a simplified protocol. For optimal results in a wider range of plasmid DNA transfection conditions, we recommend Lipofectamine™ LTX Reagent, due to its optimal balance of potency & low-cytotoxicity. Lipofectamine™ 2000 Transfection Reagent is recommended if you are transfecting a broad range of cell lines or want to transfect RNA.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 4 vial (1 mL each) of Lipofectamine™ Transfection Reagent. Store at 2°C to 8°C. Do not freeze.
Cell Type Established Cell Lines, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 4 mL
Serum Compatible No
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.