missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectin™ Transfection Reagent

Product Code. 10104432
Click to view available options
Quantity:
1 mL
4 mL
Unit Size:
1mL
4mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10104432

Brand: Invitrogen™ 18292037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains one vial (1mL) of Lipofectin Transfection Reagent.

Ideal reagent for transfection of endothelial cells

  • For transfecting DNA, RNA, and oligonucleotides into mammalian cells, and DNA into plant protoplasts
  • Lipofectin reagent has also been shown to work well, in combination with PLUS™ Reagent, for transfection of HeLa cells
  • Lipofectin Transfection Reagent is a 1:1 (w/w) liposome formulation of cationic lipid N-[1-(2,3-dioleyloxy)propyl]-n,n,n-trimethylammonium chloride (DOTMA) and dioleoyl phophotidylethanolamine (DOPE) in membrane-filtered water
  • Animal-free origin

Drug Discovery & Development, Gene Expression Analysis & Genotyping, PCR & Real-Time PCR, PCR-Based Drug Discovery Assays, Real Time PCR (qPCR), Real Time PCR-Based Gene Expression Profiling, Target & Lead Identification & Validation

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each unit contains one or multiple vials (1 ml) of reagent. Store at +4°C. Lipofectin™ Transfection Reagent is guaranteed stable for 6 months when stored properly.
Cell Type Established Cell Lines, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Includes Four vials (1mL each) of Lipofectin Transfection Reagent
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Lipofectin
Product Type Transfection Reagent
Quantity 4 mL
Serum Compatible No
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.