missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipofectin™ Transfection Reagent
Ideal reagent for transfection of endothelial cells
Brand: Invitrogen™ 18292011
349.32 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: Contains one vial (1mL) of Lipofectin Transfection Reagent.
Description
- For transfecting DNA, RNA, and oligonucleotides into mammalian cells, and DNA into plant protoplasts
- Lipofectin reagent has also been shown to work well, in combination with PLUS™ Reagent, for transfection of HeLa cells
- Lipofectin Transfection Reagent is a 1:1 (w/w) liposome formulation of cationic lipid N-[1-(2,3-dioleyloxy)propyl]-n,n,n-trimethylammonium chloride (DOTMA) and dioleoyl phophotidylethanolamine (DOPE) in membrane-filtered water
- Animal-free origin
Drug Discovery & Development, Gene Expression Analysis & Genotyping, PCR & Real-Time PCR, PCR-Based Drug Discovery Assays, Real Time PCR (qPCR), Real Time PCR-Based Gene Expression Profiling, Target & Lead Identification & Validation
Order Info
Shipping Condition: Wet Ice
Specifications
Contains one vial (1 ml) of Lipofectin™ Transfection Reagent. Store at 4°C. Do not freeze. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
One vial (1mL) of Lipofectin Transfection Reagent | |
Lipofectin™ | |
1 mL | |
Wet Ice |
Established Cell Lines, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA | |
Transfection | |
Not High-throughput Compatible (Manual) | |
Transfection Reagent | |
No |
For Research Use Only. Not for use in diagnostic procedures.