Learn More
Invitrogen™ Lysis Buffer (R2 )
Component in the PureLink 96 Plasmid Purification System
Brand: Invitrogen™ 12057022
Code : 50
Additional Details : Weight : 0.07350kg
Includes: 1L bottle
Description
- Lysis Buffer (R2) offers unique approach to high throughput plasmid DNA isolation in the PureLink 96 Plasmid Purification System
Automated Nucleic Acid Purification, Automated Plasmid DNA Purification, DNA & RNA Purification & Analysis, DNA Extraction, Miniprep, Plasmid DNA Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Lysis Buffer | |
1 L |
For Research Use Only. Not for use in diagnostic procedures.