missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ Mach1™ T1 Phage-Resistant Chemically Competent E. coli

Product Code. 10512673
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10512673

Brand: Invitrogen™ C862003

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

One Shot Mach1 T1 Phage-Resistant Chemically Competent E. coli are the fastest growing chemically competent cells with a doubling time approximately 50 minutes.

One Shot Mach1 T1 Phage-Resistant Chemically Competent E. coli are the fastest growing chemically competent cells with a doubling time approximately 50 minutes. Mach1 T1R colonies are clearly visible within eight hours of plating the transformation mix (ampicillin selection), enabling you to plate and pick colonies the same day. Plasmid DNA can be prepared four hours after inoculating a single, overnight-grown colony in the selective media of choice (with any antibiotic). Mach1 T1R Chemically Competent E. coli cells can achieve transformation efficiency of >1 x 109 cfu/μg plasmid DNA.

The Mach1 T1R strain is derived from a non-K-12, wild-type W E. coli strain (ATCC #9637) that had been selected due it’s growth speed and genetically manipulated to possess the most useful cloning strain features. The strain was engineered to carry the tonA (also known as fhuA) mutation that confers resistance to T1 and T5 phage infection. The tonA genotype offers added security to valuable clones and libraries. The Mach1 T1R strain has the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. It also carries the recA1398 mutation that helps reduce the rate of recombination while propagating plasmid DNA, the endA1 mutation that increases plasmid DNA yield and quantity, and hsdR restriction system deletion to allow genomic DNA cloning.

One Shot Mach1 T1 Phage-Resistant Chemically Competent E. coli offer:
• Transformation efficiencies of >1 x 109 cfu/μg plasmid DNA
tonA (fhuA) genotype to confer resistance to T1 and T5 phage 
lacZ∆M15 for blue-white color screening of recombinants
hsdR mutation for efficient transformation of unmethylated DNA from PCR applications
recA1398 mutation for reduced occurrence of homologous recombination in cloned DNA
endA1 mutation for increased plasmid yield and quality

Easy-to-use One Shot format 
Mach1 T1 Phage-Resistant Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, helping save time and prevent contamination.

Genotype
F φ80lacZΔM15 ΔlacX74 hsdR(rK–, mK+) ΔrecA1398 endA1 tonA

Find the strain and format that you need 
• We offer other strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• Mach1 T1R and other strains are available in MultiShot formats for high throughput applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot Mach1 T1R Chemically Competent E. coli (21 x 50 μL); store at –80°C
• pUC19 DNA (50 μL at 10 pg/μL); store at –20°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (wild-type W)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.