missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagicMedia™ E. coli Expression Medium

Product Code. 10412313
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10412313

Brand: Invitrogen™ K6803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contain proprietary component A (in SoluPouchpowder or 1X liquid format) and component B (in buffer format).

Specifically designed to dramatically increase the yield of recombinant proteins in T7-regulated E. coliexpression systems without monitoring culture O.D. (optical densities) or adding inducer

  • The proprietary formulation enables E. coligrowth to reach culture densities 3 to 10-fold higher than those achieved with traditional LB + IPTG methods
  • Achieve 3 to 10 times higher protein yields with much less hands-on time
  • Eliminate time-consuming O.D. monitoring steps: inoculate prepared MagicMediaE. coliExpression Medium, grow the culture overnight, and harvest
  • Reduce culture volume to simplify handling and scale-up processes
  • Excellent for simultaneous expression of multiple clones for high-throughput applications

Bioproduction, Cloning, Transformation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type E. Coli Expression Medium
Content And Storage Component A (1X liquid) and Component B (liquid)

Store in the dark at 2−8°C.

Shelf life: 12 months from date of manufacture
Shipping Condition Room Temperature
Target Organism Class E. coli
Quantity 1 L
Product Line MagicMedia

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.