missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Cot-1 DNA

Product Code. 11578906
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11578906

Brand: Invitrogen™ 18440016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Mouse Cot-1 DNA™ is supplied at 1mg/mL in 10mM Tris-HCl (pH 7.4), 1mM EDTA.

Mouse DNA predominantly 50 to 300 bp in size and enriched for repetitive DNA sequences such as the B1, B2, and L1 family members

Mouse Cot-1 DNA™ is commonly used to block non-specific hybridization in microarray screening. It can also be used to suppress hybridization of rodent repetitive DNA sequences when mapping rodent clones by in situ hybridization or Southern blotting and to identify mouse clones in screens of somatic-cell hybrid libraries derived from mouse-hamster hybrid cells (1-3). The amount supplied is sufficient for 5 to 10 Southerns or 500 in situ hybridizations.

Performance and Quality Testing:
Purity and DNA size are verified by agarose gel electrophoresis. Concentration is verified spectrophotometrically by diluting Mouse Cot-1 DNA™ 1:100 in 50mM NaOH and using the conversion factor 0.033μg/μL/A 260 . Other methods used to determine concentration may yield varying results.

Array CGH, Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, In Situ Hybridization (ISH), Microarray Analysis

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse DNA
Content And Storage Mouse Cot-1 DNA™ is supplied at 1 mg/ml in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA. Store at -20°C.
Reagent Type Cot-1 DNA
Shipping Condition Approved for shipment on Wet or Dry Ice
Species Mouse
Product Line Cot-1 DNA
For Use With (Application) Block Non-Specific Hybridization
Quantity 500 μg
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.