missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiSite Gateway™ Pro Plus, for flexible cloning of up to four DNA fragments into a Gateway™ destination vector

Product Code. 10312203
Click to view available options
Quantity:
20 reactions
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10312203

Brand: Invitrogen™ 12537100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Include vectors and primers, Mach1 T1R Chemically Competent Cells, and LR Clonase II Plus and BP Clonase II enzymes.

For flexible cloning of up to four DNA fragments into Gateway destination vector

  • Using specifically designed att sites for recombinational cloning, clone two, three, or four DNA fragments into any Gateway Destination vector containing attR1 and attR2 sites
  • Resulting expression clone is ready for downstream expression and analysis applications
  • Clone multiple DNA fragments into one vector without using restriction enzymes or ligases
  • Full advantage of wide selection of Gateway Destination vectors or create vectors
  • Replace multiple plasmid transfections with single vector carrying all DNA elements of interest
  • MultiSite Gateway Pro Kits are available for cloning two, three, or four DNA fragments and for flexible and customized configuration of DNA fragments into a Gateway Destination vector

Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Multi-Gene Expression, Multisite Gateway, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Promoter None (Promoterless)
Product Type Gateway System Destination Expression Vector
Content And Storage The MultiSite Gateway™ Pro Kits include vectors and primers as indicated in Table 1, One Shot™ Mach1™ T1R Chemically Competent Cells, and LR Clonase™ II Plus and BP Clonase™ II enzymes. Store cells at -80°C. Store Clonase™ enzymes at -20°C for up to 6 months, or at -80°C for long-term storage. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Cloning Method Gateway
Quantity 20 reactions
Vector MultiSite Gateway Vectors
Product Line Gateway
For Use With (Application) Clone your own Promoter, Multi-Gene Expression
compliance-icons
Product Identifier
  • MultiSite Gateway Pro Plus, for flexible cloning of up to four DNA fragments into a Gateway destination vector
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase, tritirachium album serine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.