missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ BL21(DE3) Chemically Competent E. coli

Product Code. 10749734
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
20 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10749734

Brand: Invitrogen™ C600003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One Shot BL21(DE3) E.coli: 20 vials, 50μL each (total of 1mL), pUC19 DNA (10pg/μL): 1 vial, 50μL, SOC Medium: 1 bottle, 6mL

One Shot BL21(DE3) Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET.

One Shot BL21(DE3) Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET. This strain had been engineered to carry the λDE3 lysogen that contains T7 RNA polymerase gene under control of the lacUV5 promoter. Recombinant proteins that are nontoxic to E. coli are generally expressed at higher levels in BL21(DE3) cells compared to other strains with tighter expression control.

The BL21(DE3) E. coli strain is one of the most popular host strains to produce recombinant proteins and adapted in research or industrial applications. It is a derivative of the E. coli B strain that does not contain the lon protease and is also deficient in the outer membrane protease OmpT. The lack of two key proteases reduces degradation of heterologous proteins expressed in the strains. Additionally, several other strain features make them especially suited for protein production, namely fast growth in minimal medium, a lower acetate production when grown in high glucose media, and the ability to reach high cell density. One Shot BL21(DE3) Chemically Competent E. coli cells can achieve transformation efficiency of >1 x 108 cfu/μg of control DNA.

Notes
• The basal expression levels of heterologous genes are significantly higher in BL21(DE3), thus strains such as BL21 AI are recommended for toxic protein expression.
• IPTG is required to induce expression of the T7 RNA polymerase cascade system that in turn transcribe T7 promoter regulated target gene.

One Shot BL21(DE3) Chemically Competent E. coli offer:
• Transformation efficiency of >1 x 108 cfu/μg control plasmid DNA 
• Ion and OmpT protease deficiency reduces degradation of recombinant protein 
• IPTG-inducible lacUV5 promoter controls expression of the T7 RNA polymerase 
hsdS mutation allows efficient transformation of unmethylated DNA 
• Fast growth in minimal medium and ability to reach high cell density

Easy-to-use One Shot format 
BL21(DE3) Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype: F–ompT hsdSB (rB–, mB–) gal dcm (DE3)

Find the strain and format that you need
• We offer other E. coli strains for protein expression. For expression of toxic proteins, consider BL21-AI™ One Shot™ Chemically Competent E. coli (Cat. No. C607003).
• BL21 Star (DE3) and other strains are available in MultiShot formats for high throughput applications.
• We offer a variety of systems for the expression of recombinant proteins in E. coli., the Champion pET expression system provides the highest level of protein production available in any expression system.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality No
Cloning Methylated DNA No
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • One Shot BL21(DE3) Chemically Competent E.coli (21 x 50 μL); store at –80°C
• pUC19 DNA (1 x 50 μL); store at –20°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial No
Blue/White Screening No
High-throughput Compatibility Low
Preparing Unmethylated DNA Yes (dcm)
Reduces Recombination No
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (B)
Promoter T7
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Toxic Proteins No
Improves RNA Stability No
Improves Protein Stability Yes (lon, ompT)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.