missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ MAX Efficiency™ DH5α-T1R Competent Cells

Product Code. 10377742
Click to view available options
Quantity:
21 x 50 μL
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10377742

Brand: Invitrogen™ 12297016

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One Shot MAX Efficiency DH5α-T1R Competent Cells: 20 vials, 50μL each ( total of 1mL), pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 1 bottle, 6mL

One Shot MAX Efficiency DH5α-T1R Competent Cells are derived from the popular DH5α strain and carry the tonA (fhuA) mutation that confers resistance to T1 and T5 phage.

One Shot MAX Efficiency DH5α-T1R Competent Cells are derived from the popular DH5α strain and carry the tonA (fhuA) mutation that confers resistance to T1 and T5 phage. They offer added security to protect valuable clones and libraries. This is especially important for genome and sequencing centers where T1 phage infection would be detrimental.

DH5α-T1R is a versatile strain that can be used in many everyday cloning applications. Prepared by a proprietary modification of the Hanahan method, these cells are suitable for the construction of gene banks or for the generation of cDNA libraries using plasmid-derived vectors. The lacZ∆M15 marker provides α-complementation of the ß-galactosidase gene from pUC or similar vectors and therefore can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α-T1R Competent Cells are capable of being transformed efficiently with large plasmids and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF'IQ is provided to allow plaque formation.

Benefits of MAX Efficiency DH5α-T1R Competent Cells
• Transformation efficiency of >1 x 109 transformants/μg with non-saturating amounts of DNA
tonA (fhuA) mutation to confer resistance to T1 and T5 phage
endA1 mutation that enables cleaner DNA preparations and better results in downstream applications due to the elimination of nonspecific digestion by Endonuclease I
recA1 mutation that reduces occurrence of unwanted recombination in cloned DNA
hsdR for efficient transformation of unmethylated DNA from PCR applications  
lacZΔM15 marker that allows blue/white color screening of recombinant clones carrying the alpha fragment of β-galactosidase 
 
Convenient and efficient format 
DH5α-T1R E. coli are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation so there are no efficiency-zapping, freeze-thaw cycles or money wasted on unused cells.

Genotype: F- φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK-, mK+) phoA supE44 thi-1 gyrA96 relA1 tonA

Find the strain and format that fits you needs
• DH strains are available in chemically competent and electrocompetent cell formats.
• MAX Efficiency DH5αF´IQ Competent Cells (Cat. No. 18288019) are available in chemically competent format.
• DH5α-T1R is available in Multishot format for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot MAX Efficiency DH5α-T1R Competent Cells (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.