Learn More
Invitrogen™ One Shot™ PIR1 Chemically Competent E. coli
One Shot PIR1 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replication (e.g., pUni/V5-His-TOPO for recombination with an Echo cloning system).
Brand: Invitrogen™ C101010
283.10 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Includes: One Shot PIR1 Chemically Competent E. colicontains ten transformations of chemically competent cells in One Shot format.
Description
One Shot PIR1 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replication (e.g., pUni/V5-His-TOPO for recombination with an Echo cloning system). The pir gene encodes the replication protein π, which is required to replicate and maintain plasmids containing the R6Kγ origin. The PIR1 strain contains a mutant allele of the pir gene that maintains the donor vector construct at ∼250 copies per cell. PIR1 cells are competent and can achieve high transformation efficiency of >1 x 109 cfu/μg supercoiled plasmid.
R6Kγ allows tight control and selection and has been used in various applications including genome engineering, mutagenesis studies, bacterial artificial chromosome (BAC) construction, high-capacity extrachromosomal vector systems, and suicide plasmid construction.
One Shot PIR1 Chemically Competent E. coli cells offer:
• pir gene to maintain vectors that contain the R6Kγ ori at ∼250 copies per cell
• Transformation efficiencies of >1 x 109 cfu/μg
• endA1 mutation enables increased plasmid DNA yield and quantity
• recA1 mutation reduce occurrence of unwanted recombination in cloned DNA
• hsdR for efficient transformation of unmethylated DNA from PCR applications
Convenient and efficient format
PIR1 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation, reducing freeze-thaw cycles and money wasted on unused cells.
Genotype
F–∆lac169 rpoS(Am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir-116
Genetic marker descriptions
Find the strain and format that fit your needs
• Multiple DH strains are available in chemically competent and electrocompetent cell formats.
• We offer PIR2 strain (Cat. No. C111110) that carries wild-type pir gene and maintain vectors at ∼15 copies per cell.
• MultiShot formats are available for high throughput applications.
Specifications
Chemically Competent Cells | |
Yes (endA1) | |
High Efficiency (>1 x 109 cfu/μg) | |
No | |
Low | |
Yes (recA1) | |
No | |
Tube | |
11 x 50 μL |
No | |
No | |
• One Shot PIR1 Chemically Competent E. coli (11 x 50 μL) Store Competent Cells at –80°C. • pUC19 plasmid (50 μL at 10 pg/μL) Store pUC19 plasmid at –20°C. • S.O.C. Medium Store S.O.C. Medium at 4°C or room temperature. |
|
No | |
No | |
Dry Ice | |
E. coli | |
One Shot™ |
For Research Use Only. Not for use in diagnostic procedures.