missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ Stbl3™ Chemically Competent E. coli

Product Code. 10193952
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10193952

Brand: Invitrogen™ C737303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One Shot Stbl3 Competent E. coli: 21 vials, 50μL each, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 1 bottle, 6mL

One Shot Stbl3 Chemically Competent E. coli are designed for cloning direct repeats found in lentiviral expression vectors as these cells reduce the frequency of homologous recombination of direct repeats found in lentiviral vectors (ViraPower Lentiviral Expression kits) and other retroviral vectors (instability-prone HIV-based plasmids).

One Shot Stbl3 Chemically Competent E. coli are designed for cloning direct repeats found in lentiviral expression vectors as these cells reduce the frequency of homologous recombination of direct repeats found in lentiviral vectors (ViraPower Lentiviral Expression kits) and other retroviral vectors (instability-prone HIV-based plasmids). The Stbl3 strain has been essential for many applications, including genome editing CRISPR workflows, vaccine development (including SARS-CoV-2 vaccine research), cloning, and maintaining vectors with multiple repeats (e.g., gene coding protein with repeated sequence). The transformation efficiency of One Shot Stbl3 chemically competent cells is >1 x 108 cfu/μg plasmid DNA.

The Stbl3 strain is derived from the HB101 E. coli strain with a hybrid K-12 and B strain genome. It contains the recA13 allele of the recA mutation that reduces nonspecific recombination occurrence in cloned DNA. Mutations in the Stbl3 methylation-dependent restriction system (mcrB, mrr, and hsdS20) allow construction of cloning genomic DNA. This strain has endA1+, a nonspecific endonuclease I that digests DNA. The Stbl3 strain also reproducibly yields more plasmid DNA than DH strains (10-fold or more plamsmid DNA than the TOP10 strain) and sustainably produces high titer lentivirus.

One Shot Stbl3 Chemically Competent E. coli offer:
• Expected efficiency of >1 x 108 transformants/μg plasmid DNA
• Optimal performance for the propagation of lentiviral vectors and maintenance of direct repeats
• Enhanced genomic DNA cloning capacity (mcrB, mrr, and hsdS20)
• Capable of producing high DNA yields 

Note: For optimal performance and to maintain plasmid with complex repeat sequences, Stbl3 cells should be cultured at 30°C. When performing plasmid DNA isolation from Stbl3 cells that have wild type endonuclease I (endA1+), ensure that buffers contain 10 mM EDTA. EDTA will inactivate the endonuclease and avoid DNA nicking and vector degradation (follow plasmid DNA purification kit instructions for endA1+ E. coli strains).

Easy-to-use One Shot format
Stbl3 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype
FmcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl-1

Find the strain and format that fit your needs
• Other Stbl strains are available with different genetic background.
• The Stbl3 strain is available in MultiShot format for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality No
Cloning Methylated DNA Yes (mcrB, mrr)
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • One Shot Stbl3 Competent E. coli (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

.
Antibiotic Resistance Bacterial (Yes) Streptomycin
Cloning Unstable DNA Yes (recA13)
Blue/White Screening No
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA13)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12/B hybrid)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.