missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pBAD/Myc-His Kit

Expresses native proteins or fusion proteins with a C-terminal tag in a tightly regulated fashion

Brand:  Invitrogen™ V44001

Code : 50

Additional Details : Weight : 0.14000kg

Product Code. 10580333

  • £932.00 / 1 set
Estimated Shipment: 29-07-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Includes: 20μg of pBAD/Myc-His A, B and C vector, 20μg of pBAD/Myc-His/lacZ, 20% L-arabinose, LMG194 and TOP10 E. coli stabs.

Description

Description

  • araBAD promoter for tightly regulated expression
  • Translation initiation signals optimized for E. coli expression
  • C-terminal polyhistidine (6xHis) tag for purification with nickel-chelating resin or detection with Anti-His(C-term) Antibody
  • C-terminal c-myc epitope for detection and analysis with Anti-myc Antibody
  • Three vectors are provided (A, B, and C): Each has C-terminal tag in different reading frame relative to multiple cloning site to simplify in-frame cloning of gene

Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Specifications

Specifications

Inducible
araBAD
None
Ampicillin (AmpR)
c-Myc Epitope Tag
pBAD
Arabinose
Bacterial Expression Vector
The pBAD/Myc-His Kit includes 20 μg of pBAD/Myc-His A, B, & C vector, 20 μg of pBAD/Myc-His/lacZ, 20% L-arabinose, LMG194 and TOP10 E. coli stabs. Vectors and 20% L-arabinose should be stored at -20°C. Store LMG194 and TOP10 stabs at 2 - 8°C. All components are guaranteed stable for 6 months when properly stored.
EK (Enterokinase) Recognition Site
Restriction Enzyme/MCS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.