Learn More
Invitrogen™ pBAD TOPO™ TA Expression Kit
Specifically designed for one-step cloning and regulated prokaryotic expression of Taq-amplified PCR products
Brand: Invitrogen™ K430040
1313.82 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Description
- Linearized, topoisomerase I-activated vector for 5-minute cloning of Taqamplified PCR products
- araBAD promoter for tightly regulated expression in E. coli V5 epitope tag for detection with Anti-V5 Antibody
- C-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-His(C-term) Antibody
- Enterokinase cleavage site for removal of N-terminal leader peptide
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
araBAD | |
None | |
Ampicillin (AmpR) | |
EK (Enterokinase) Recognition Site | |
Cell-Based Expression | |
TOPO-TA | |
pBAD |
Arabinose | |
TA Expression Kit | |
Each pBAD TOPO™ TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO™ TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200 ng of topoisomerase I-activated pBAD-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot™ box contains all of the reagents necessary for transformation, including single-use 50 μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ cells at -80°C. Store the LMG194 E. coli stab at at 2 - 8°C. All reagents are guaranteed stable for 6 months when properly stored. | |
LMG194, TOP10 | |
E. coli | |
V5 Epitope Tag | |
40 Reactions | |
TOPO™ |
For Research Use Only. Not for use in diagnostic procedures.