Learn More
Invitrogen™ pBAD202 Directional TOPO™ Expression Kit
Clones PCR product at high efficiency in five minutes into a topoisomerase I-activated expression vector
Brand: Invitrogen™ K420201
Code : 50
Additional Details : Weight : 0.00500kg
Description
Using restriction enzymes to clone gene into an expression vector often forces to compromise final sequence of insert, especially when there are no useful restriction sites close to genes coding sequence. This may result in suboptimal spacing of expression elements or incorporation of non-native amino acid residues, which can reduce expression levels and/or cause production of non-functional protein.
- TOPO Expression Vectors enable to insert exact DNA sequence require simply by performing PCR with appropriately designed primers
- Resulting recombinant expression vector contains exact DNA sequence without any noncoding regions
- Several expression vectors are adapted for Directional TOPO Cloning, enabling to use proofreading polymerases and to clone the PCR products in a specific orientation
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
araBAD | |
None | |
LMG194, TOP10 | |
E. coli | |
His Tag (6x), V5 Epitope Tag, Thioredoxin | |
pBAD |
Arabinose | |
TOPO Expression Kit | |
Kanamycin (KanR) | |
EK (Enterokinase) Recognition Site | |
Cell-Based Expression | |
Directional TOPO | |
TOPO™ |
For Research Use Only. Not for use in diagnostic procedures.