Learn More
Invitrogen™ pcDNA™3.1 (-) Mammalian Expression Vector
Designed for high-level, constitutive expression in a variety of mammalian cell lines
Brand: Invitrogen™ V79520
Code : 50
Additional Details : Weight : 0.02000kg
Includes: 20μg of this pcDNA3.1(-) vector, as well as an expression control, are supplied supercoiled and lyophilized.
Description
- Contains Geneticin selectable marker and forward-orientation multiple cloning site
- For use alone or in co-transfections
- Cytomegalovirus (CMV) enhancer-promoter for high-level expression
- Large multiple cloning site in reverse (-) orientation
- Bovine Growth Hormone (BGH) polyadenylation signal and transcription termination sequence for enhanced mRNA stability
- SV40 origin for episomal replication and simple vector rescue in cell lines expressing large T antigen (i.e.,COS-1 and COS-7)
- Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Transfection | |
Mammalian Expression Vector | |
20 μg of this pcDNA™3.1(-) vector, as well as an expression control, are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored. | |
Restriction Enzyme/MCS | |
pcDNA™ |
CMV | |
Geneticin™ (G-418) | |
Untagged | |
pcDNA | |
Constitutive Expression |
For Research Use Only. Not for use in diagnostic procedures.