Learn More
Invitrogen™ pcDNA™3.1/Zeo (-) Mammalian Expression Vector
Designed for high-level, constitutive expression in a variety of mammalian cell lines
Brand:  Invitrogen™ V86520
490.96 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 11550876
  • £722.00 / 20µg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Includes: 20μg of this pcDNA3.1/Zeo(-) vector, as well as an expression control, are supplied supercoiled and lyophilized.
Description
Description
  • Contains Zeocin selectable marker and reverse-orientation multiple cloning site
  • Cytomegalovirus (CMV) enhancer-promoter for high-level expression
  • Large multiple cloning site in either forward (+) or reverse (-) orientations
  • Bovine Growth Hormone (BGH) polyadenylation signal and transcription termination sequence for enhanced mRNA stability
  • SV40 origin for episomal replication and simple vector rescue in cell lines expressing the large T antigen (i.e.,COS-1 and COS-7)
  • Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Constitutive
CMV
Zeocin™
Untagged
20 μg
pcDNA™
Transfection
Mammalian Expression Vector
20 μg of this pcDNA™3.1/Zeo(-) vector, as well as an expression control, are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored.
Restriction Enzyme/MCS
pcDNA
Constitutive Expression