Learn More
Invitrogen™ pcDNA™4/TO Mammalian Expression Vector
Uses the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from an especially strong mammalian promoter sequences
Brand: Invitrogen™ V102020
Code : 50
Additional Details : Weight : 0.06000kg
Description
- Uses repressor mechanism that blocks transcription from the powerful CMV promoter in absence of tetracycline
- Because the T-REx System elements do not use viral transactivators, user can achieve high-level expression from complete CMV promoter without secondary, nonspecific activation of host genes
- Two tetracycline operator sequences (TetO2) have been inserted between TATA box of the CMV promoter and the transcriptional start site: The TetO2 sequence itself has no effect on expression When tetracycline repressor protein (TR) is present, it effectively binds TetO2 sites and blocks transcription initiation
- Tetracycline added to culture medium binds to, and changes conformation of, TR protein
- This change causes TR protein to release TetO2 sites, derepressing transcription from the CMV promoter for high-level expression of gene of interest
- Expression levels can be modulated based on tetracycline concentration and can be induced to levels that are achieved with constitutive CMV expression vectors
- Zeocin or hygromycin resistance gene for effective selection of stable mammalian cell lines
- Large multiple cloning site to simplify cloning
Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Targeted Integration
Specifications
Inducible | |
Tetracycline | |
Mammalian Expression Vector | |
Untagged | |
pcDNA | |
Regulated Expression |
Transfection | |
CMV/TO | |
Zeocin™ | |
Restriction Enzyme/MCS | |
T-REx™, pcDNA™ |
For Research Use Only. Not for use in diagnostic procedures.