missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pcDNA™6/TR vector Mammalian Expression Vector

Product Code. 11550806
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11550806

Brand: Invitrogen™ V102520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

20μg of supercoiled vector (40μL at 0.5μg/μL) supplied in TE Buffer, pH 8.0.

Uses the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from this strong mammalian promoter sequences

  • Regulatory vector, pcDNA6/TR, is provided for high-level expression of tetracycline repressor (TR) protein
  • This vector expresses Blasticidin resistance gene for rapid selection of mammalian cell lines that stably express TR protein

Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Targeted Integration

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Promoter CMV
Product Type Mammalian Expression Vector
Selection Agent (Eukaryotic) Blasticidin
Content And Storage 20 μg of supercoiled vector (40 μL at 0.5 μg/μL) supplied in TE Buffer, pH 8.0. Store at -20°C.
Shipping Condition Room Temperature
Protein Tag Untagged
Quantity 20 μg
Vector pcDNA
Product Line T-REx, pcDNA
For Use With (Application) Regulated Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.