Learn More
Invitrogen™ pCEP4 Mammalian Expression Vector
Designed for high-level, constitutive expression from either the CMV or RSV promoters
Brand: Invitrogen™ V04450
Code : 50
Additional Details : Weight : 0.00450kg
Includes: 20μg of each vector is supplied supercoiled and lyophilized.
Description
- Contain EBNA-1 gene for episomal expression in primate and canine cell lines
Antibody and Cell Culture Production, Bioproduction, Constitutive Expression, Episomal Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Stable Cell Line Development
Specifications
Constitutive | |
CMV | |
Hygromycin | |
Untagged | |
pCEP4 |
Transfection | |
Mammalian Expression Vector | |
20 μg of pCEP4 vector in Tris-EDTA buffer Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored. |
|
Restriction Enzyme/MCS | |
Stable Cell Line Development, Episomal Expression |
For Research Use Only. Not for use in diagnostic procedures.