missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCRX Enhancer System

PCRx Enhancer System is used in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates.

Brand:  Invitrogen™ 11495017

53.44 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.



Product Code. 10781144

  • £71.25 / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: PCRx Enhancer Solution consists of 10X PCRx Amplification Buffer, 10X PCRx Enhancer Solution, and 50mM magnesium sulfate.
This item is not returnable. View return policy

Description

Description

PCRX Enhancer System is an optimized buffer and cosolvent system designed for use in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates. 10X PCRX Enhancer Solution with 10X PCRX Amplification Buffer offers higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostability of DNA polymerases such as Taq DNA polymerase and Platinum Taq DNA Polymerase.

Conventional PCR buffer components can be directly substituted with the 10X PCRX amplification buffer and 50 mM MgSO4 for a more robust amplification and improved product yield in some cases.

Notes

Optimal concentration of 10X PCRX Enhancer Solution will vary depending on GC content, Mg++ concentration, and annealing temperature.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10X PCRx Enhancer Solution (2 x 1 mL)
• 10X PCRx Amplification Buffer (2 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -20°C.
250 rxns
Standard PCR, Hot-start PCR
Separate Components
Platinum Taq DNA Polymerase, Platinum Taq DNA Polymerase High Fidelity, Taq DNA Polymerase
Dry Ice
250 Reactions
DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PCRX Enhancer System

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 4-methylmorpholine-4-oxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.